Learn More
Munktell Filtrak™ Micro-Glass Fiber Filters
Achieve optimum filtration. Munktell Filtrak™ Micro-Glass Fibers used in Munktell filters are made of 100% borosilicate glass of varying diameters fine-tuned to the particle retention and flow requirements of the grades.
Brand:  Munktell Filtrak™ 410032
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11387274
  • 107.00 EUR / Pack of 100
Estimated Shipment: 18-04-2024
to see stock.
Add to basket
Description
Description
  • Binder-free micro-glass fiber filters
  • Micro-glass fiber filters with binders
  • Micro-glass fiber filters, supported
Specifications
Specifications
Borosilicate Glass
Solvent filtration, Scintillation counting, Highly efficient for general laboratory filtration, clarification of buffer and reagent solutions, corresponds to many international standards for air and water pollution monitoring. Foodstuff analysis, filtration of algae, bacteria cultures, proteins
0.23 mm
1.6 μm
MGA
110 mm