Learn More
Munktell Filtrak™ Micro-Glass Fiber Filters
Achieve optimum filtration. Munktell Filtrak™ Micro-Glass Fibers used in Munktell filters are made of 100% borosilicate glass of varying diameters fine-tuned to the particle retention and flow requirements of the grades.
35.00 EUR - 107.00 EUR
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11387274
|
Munktell Filtrak™
410032 |
110 mm |
107.00 EUR
Pack of 100 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11327304
|
Munktell Filtrak™
410005 |
90 mm |
35.00 EUR
Pack of 50 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
Description
- Binder-free micro-glass fiber filters
- Micro-glass fiber filters with binders
- Micro-glass fiber filters, supported