Learn More
Munktell Filtrak™ Micro-Glass Fiber Filters
Achieve optimum filtration. Munktell Filtrak™ Micro-Glass Fibers used in Munktell filters are made of 100% borosilicate glass of varying diameters fine-tuned to the particle retention and flow requirements of the grades.
Brand: Munktell Filtrak™ 410032
Additional Details : Weight : 0.01000kg
Description
- Binder-free micro-glass fiber filters
- Micro-glass fiber filters with binders
- Micro-glass fiber filters, supported
Specifications
Borosilicate Glass | |
Solvent filtration, Scintillation counting, Highly efficient for general laboratory filtration, clarification of buffer and reagent solutions, corresponds to many international standards for air and water pollution monitoring. Foodstuff analysis, filtration of algae, bacteria cultures, proteins | |
0.23 mm |
1.6 μm | |
MGA | |
110 mm |