Learn More
Eppendorf™ Micro Fusion Chamber
Easy to use. Integrated cuvette chamber and extremely high transformation rates
Brand: Eppendorf™ 4308030003
Additional Details : Weight : 0.09000kg
Description
- Extremely high transformation rates due to innovative electronics and buffers
- Integrated cuvette chamber
- Easy to use
- Compact design
- Universal electroporator for eukaryotic, yeast and bacteria
Specifications
For Fusion | |
1 Ea. | |
0.2 mm |
Micro Fusion Chamber | |
1 |