Learn More
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Transform your laboratory or workplace storage as you see fit. Metro Super Erecta Wire Shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems
Brand:  Metro™ 2160NS
Additional Details : Weight : 6.71200kg
 View more versions of this product
Product Code. 15956641
  • 560.00 EUR / Each
Estimated Shipment: 12-09-2024
Description
  • Open-wire design minimizes dust accumulation, allows free circulatin of air and greater visibility of stored items
  • Type 304 polished stainless steel
  • Used for the most aggressive applications and environments
  • Stands up to high temperature automated cart wash and autoclave applications
Specifications
Specifications
53 x 152.4 in. (21 x 60 cm)
21 in.
Metro™ Super Erecta™ shelving system
Plastic sleeves and wedges
530 mm