Learn More
Metro™ Double-Deep qwikTRAK™ Stationary Intermediate Unit Kits
Connects one double-deep stationary imtermediate unit to to tracks and anchors tracks to floor
Brand: Metro™ LBTAK3
Additional Details : Weight : 14.51200kg
Description
- For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ and MetroMax iQ shelving systems
- Kit includes 74 in. (188cm) high stationary posts and connecting hardware
Order Info
Shelves are sold separately.
Specifications
74 in. High stationary posts and connecting hardware to connect double-deep qwikTRAK™ stationary intermediate units to track and anchors track to floor | |
Stationary Intermediate Unit Kit | |
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ shelving system | |
Steel |
18 kg | |
39.6 lb. | |
Metroseal Green Epoxy |