Learn More
Metro™ Double-Deep qwikTRAK™ Stationary End Unit Kits
Connects end units to tracks and anchors tracks to floor
1265.00 EUR - 2370.00 EUR
Specifications
Includes | 74 in. High stationary posts and connecting hardware to connect double-deep qwikTRAK™ stationary end units to track and anchors track to floor |
---|---|
Product Type | Stationary End Unit Kit |
Product Code | Brand | For Use With (Equipment) | Material | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Material | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
15936981
|
Metro™
LBTES |
Metro™ Super Erecta™, Super Adjustable™ shelving system | Stainless Steel | 73.6 lb. | 33.5 kg |
2370.00 EUR
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15916981
|
Metro™
LBTEC |
Metro™ Super Erecta™, Super Adjustable™ shelving system | Chrome | 73.6 lb. | 33.5 kg |
1265.00 EUR
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15926981
|
Metro™
LBTEK3 |
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ shelving system | Steel | 73.6 lb. | 33.5 kg |
1515.00 EUR
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15946981
|
Metro™
LBTEX3 |
Metro™ MetroMax i™ and Q™ shelving system | Polymer | 42.3 lb. | 19.2 kg |
2025.00 EUR
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ and MetroMax iQ shelving systems
- Kit includes 74 in. (188cm) high stationary posts and connecting hardware
Order Info
Order one kit per double-deep system. Shelves are sold separately.
Specifications
74 in. High stationary posts and connecting hardware to connect double-deep qwikTRAK™ stationary end units to track and anchors track to floor | |
Stationary End Unit Kit |