Learn More
Metro™ Cage Rack Covers, Autoclavable
Sized to fit Metro autoclavable cage racks
Brand:  Metro™ LC2460AC
Additional Details : Weight : 2.54400kg
 View more versions of this product
Product Code. 15937011
  • 860.00 EUR / Each
Estimated Shipment: 10-07-2024
Description
  • Rated for approximately 70 cycles per cover
Specifications
Specifications
Cart Cover
62 x 26 x 61.5 in. (157.5 x 66 x 156.2 cm)
61.5 in.
62 in.
26 in.
Autoclavable
24 in. W Cage rack
156.2 cm
157.5 cm
66 cm