Learn More
alpha-Methylhydrocinnamic acid, 98%, Thermo Scientific Chemicals
Catalog Number p-3212360
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
1009-67-2
C10H12O2
164.20
MFCD00192301
MCIIDRLDHRQKPH-UHFFFAOYNA-N
2-benzylpropionic acid, alpha-methylhydrocinnamic acid, 2-methyl-3-phenylpropionic acid, 2-benzylpropanoic acid, 2-methyl-3-phenyl-propionic acid, acmc-20ao3h, alpha-methylhydrocinnamicacid, alpha-methyl-hydrocinnamic acid, 2-methyl-3-phenyl-propanoic acid, #
99862
2-methyl-3-phenylpropanoic acid
CC(CC1=CC=CC=C1)C(O)=O
1009-67-2
C10H12O2
164.20
MFCD00192301
MCIIDRLDHRQKPH-UHFFFAOYNA-N
2-benzylpropionic acid, alpha-methylhydrocinnamic acid, 2-methyl-3-phenylpropionic acid, 2-benzylpropanoic acid, 2-methyl-3-phenyl-propionic acid, acmc-20ao3h, alpha-methylhydrocinnamicacid, alpha-methyl-hydrocinnamic acid, 2-methyl-3-phenyl-propanoic acid, #
99862
2-methyl-3-phenylpropanoic acid
CC(CC1=CC=CC=C1)C(O)=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 41°C
Color White to Yellow
Boiling Point 167°C to 168°C (23.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2CH(CH3)CO2H
Quantity 1 g
Formula Weight 164.2
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material α-Methylhydrocinnamic acid