missing translation for 'onlineSavingsMsg'
Learn More

Methyl 7-chloroheptanoate, 98%, Thermo Scientific™

Product Code. p-7029614
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11406180

Brand: Thermo Scientific Alfa Aesar H27098.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26040-62-0
Molecular Formula C8H15ClO2
Molecular Weight (g/mol) 178.66
MDL Number MFCD09751145
InChI Key YKSFGXAJDCLJKE-UHFFFAOYSA-N
Synonym 7-chloroheptanoic acid, methyl ester, heptanoic acid, 7-chloro-, methyl ester, acmc-20akfu, methyl 7-chloroheptanoate, 7-chloroheptanoic acid methyl ester, heptanoic acid, 7-chloro, methyl ester
PubChem CID 529989
IUPAC Name methyl 7-chloroheptanoate
SMILES COC(=O)CCCCCCCl

Specifications

Linear Formula Cl(CH2)6CO2CH3
Refractive Index 1.4425
Quantity 5 g
Formula Weight 178.66
Percent Purity 98%
Chemical Name or Material Methyl 7-chloroheptanoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.