Learn More
Methyl 2-furoate, 98%, Thermo Scientific Chemicals
Catalog Number p-163277
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
611-13-2
C6H6O3
126.11
MFCD00003236
HDJLSECJEQSPKW-UHFFFAOYSA-N
methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl ester
11902
methyl furan-2-carboxylate
COC(=O)C1=CC=CO1
611-13-2
C6H6O3
126.11
MFCD00003236
HDJLSECJEQSPKW-UHFFFAOYSA-N
methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl ester
11902
methyl furan-2-carboxylate
COC(=O)C1=CC=CO1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Yellow to Brown
Density 1.1760g/mL
Boiling Point 181.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4830 to 1.4890
Quantity 25 g
Merck Index 15,6149
Specific Gravity 1.176
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 126.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 2-furoate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-furoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.