missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, ACS HPLC Grade, Multisolvent™, Scharlab
Click to view available options
Quantity:
2.5 L
Description
Multisolvent™ Methanol is microfiltered through membranes of pore diameter 0.22μm and is suitable for liquid chromatography.
- Synonyms: Methyl alcohol, Carbinol, Methynol, Wood alcohol
- Molecular Formula: CH3OH
- Molar Mass: 32.04g/mol
- CAS: 67-56-1
- EIN ECS No.: 200-659-6
- Dielectric Constant: 32.6 (at 25°C)
Specifications
Melting Point | -98°C |
Absorbance | Passes Test |
CAS | 67-56-1 |
Acidity | 0.0002mEq/g max |
Alkalinity | 0.0002mEq/g max |
Appearance | Clear |
Density | 0.79g/cm3 |
Boiling Point | 64.5°C |
Quantity | 2.5 L |
Flash Point | 11°C |
Show More |
Safety and Handling
Product Identifier
- Methanol, Multisolvent HPCL grade ACS ISO UV-VIS K.F.
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'