Learn More
OHAUS™ Media Bottle Clamp
OHAUS™ Media Bottle Clamp is designed to securely grip and position various sizes of bottles.
109.00 EUR - 118.00 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15792509
|
OHAUS™
30400097 |
500 mL Bottle |
109.00 EUR
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15702519
|
OHAUS™
30400098 |
1000 mL Bottle |
118.00 EUR
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
- Constructed of stainless steel
- For use with media bottles
- Designed to securely hold bottles in place
Specifications
Bottle Clamp | |
Stainless Steel |