Learn More
MP Biomedicals Mccollum & Davis Salt Mixture No. 186
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 kg
Unit Size:
10kg
Description
Salt mixture containing calcium lactate and potassium phosphate.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Product Type | Animal Diet |
| Disposable | Single-use |
| Quantity | 10 kg |
| Sterility | Non-sterile |
| Storage Requirements | Room Temperature |
| Length (English) | 1 ft. |
| Length (Metric) | 0.3048 m |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.