missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MAPK1 (ERK2), GST Tag Recombinant Protein
Human MAPK1 (ERK2), GST Tag Recombinant Protein
674.00 EUR - 7890.00 EUR
Specifications
Accession Number | NP_620407 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 69.7 kDa |
Gene Alias | MAPK1 (ERK2) |
KinaseFamily | MAP Kinase Family |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10296952
|
Invitrogen
PV3595 |
1mg |
7890.00 EUR
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11523427
|
Invitrogen
PV3313 |
10μg |
674.00 EUR
10µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MAPK1 (ERK2) is a serine⁄threonine kinase that acts as an important activator of p90 RSK, MSK, ELK1, and Stat3, and is involved in the MAPK pathway.Specifications
NP_620407 | |
69.7 kDa | |
MAP Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
MAPK1 (ERK2) | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: CMGC Group | |
E. coli | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
For Research Use Only. Not for use in diagnostic procedures.