missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP3K8 (COT) Recombinant Human Protein
MAP3K8 (COT) is a serine/threonine kinase.
1800.00 EUR - 7890.00 EUR
Specifications
Accession Number | NP_005195 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 69.9kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458633
|
Invitrogen™
PR7887A |
100μg |
1800.00 EUR
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10412713
|
Invitrogen™
PV4330 |
1mg |
7890.00 EUR
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MAP3K8 can activate both the MAP and JNK kinase pathways and plays a role in the cell cycle.
Specifications
NP_005195 | |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
STE Unique Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
69.9kDa | |
RUO - research use only | |
MAP3K8 (COT) | |
Human | |
GST Tag | |
Frozen |