Learn More
Macherey-Nagel™ Manual Decappers
240.00 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10276711
|
Macherey-Nagel™
735913 |
13 mm Aluminum Crimp Caps |
240.00 EUR
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
10745962
|
Macherey-Nagel™
735920 |
20 mm Aluminum Crimp Caps |
240.00 EUR
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
Specifications
Manual Decapper | |
1 Ea. |