Learn More
Fisherbrand™ Manual Decapper, Steel, Non-autoclavable
Manual decrimping tools allow easy removal of aluminum seals without breakage.
Brand: Fisherbrand™
1154.25 EUR valid until 2024-12-31
Use promo code "23262" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
- Decapper
- Manual
- Steel
Specifications
Aluminum Cap | |
32mm Decapper |