Learn More
Magnesium Chloride Hexahydrate, purum p.a, ≥98.0% (KT), Honeywell™
Catalog Number p-7120530
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly bottle
7791-18-6
Cl2H12MgO6
203.30
MFCD00149781
DHRRIBDTHFBPNG-UHFFFAOYSA-L
magnesium(2+) hexahydrate dichloride
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
7791-18-6
Cl2H12MgO6
203.30
MFCD00149781
DHRRIBDTHFBPNG-UHFFFAOYSA-L
magnesium(2+) hexahydrate dichloride
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
Purum p.a.,≥98.0%
  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY
Melting Point 117°C
Color Colorless
pH 5.0 to 6.0 (at 20°C)
Physical Form Crystalline
Quantity 1 kg
Linear Formula MgCl2 · 6H2O
UN Number NONH for all modes of transport
Formula Weight 203.3g/mol
Percent Purity ≥98.0% (KT)
Packaging Poly bottle
Chemical Name or Material Magnesium chloride hexahydrate