Learn More
Magnesium Carbonate, Pentahydrated Basic, Extra Pure, Light, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical
Brand: Fisher Chemical M/0500/53
Additional Details : CAS Number : 12125-28-9 Weight : 0.50000kg
Description
500g, Plastic powder jar
Specifications
White | |
Solid | |
CMgO3·1/4H2O2Mg·5/4H2O | |
Plastic powder jar |
12125-28-9 | |
500 g | |
149784 |