Learn More
Magnesium carbonate basic, heavy, extra pure, Thermo Scientific Chemicals
Catalog Number p-44418
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
39409-82-0
C4H10Mg5O18
467.63
MFCD00149783
YMKSTRLFWJQKFP-UHFFFAOYSA-D
pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
39409-82-0
C4H10Mg5O18
467.63
MFCD00149783
YMKSTRLFWJQKFP-UHFFFAOYSA-D
pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Magnesium carbonate basic, heavy, 40 to 45% (MgO), CMgO3, CAS Number-39409-82-0, 500g, loss on calcination: 55 to 60%, White, 208-915-9, 84.31, Extra Pure, MgCO3, 15,5724, MFCD00011107, Plastic bottle, Powder, OM2470000
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 500 g
Linear Formula MgCO3
Merck Index 15,5724
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol, soluble in acids
Formula Weight 84.31
Percent Purity 40 to 45% (MgO)
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Magnesium carbonate basic, heavy