missing translation for 'onlineSavingsMsg'
Learn More

Lycopene, >90%, MP Biomedicals™

Catalog Number p-4405874
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy
502-65-8
C40H56
536.888
OAIJSZIZWZSQBC-GYZMGTAESA-N
lycopene, psi,psi-carotene, all-trans-lycopene, trans-lycopene, lycopene 7, lycored, lycopene van, lycopene, all-trans, unii-sb0n2n0wv6, ccris 7925
446925
CHEBI:15948
(6E,8E,10E,12E,14E,16E,18E,20E,22E,24E,26E)-2,6,10,14,19,23,27,31-octamethyldotriaconta-2,6,8,10,12,14,16,18,20,22,24,26,30-tridecaene
CC(=CCCC(=CC=CC(=CC=CC(=CC=CC=C(C)C=CC=C(C)C=CC=C(C)CCC=C(C)C)C)C)C)C
This item is not returnable. View return policy
502-65-8
C40H56
536.888
OAIJSZIZWZSQBC-GYZMGTAESA-N
lycopene, psi,psi-carotene, all-trans-lycopene, trans-lycopene, lycopene 7, lycored, lycopene van, lycopene, all-trans, unii-sb0n2n0wv6, ccris 7925
446925
CHEBI:15948
(6E,8E,10E,12E,14E,16E,18E,20E,22E,24E,26E)-2,6,10,14,19,23,27,31-octamethyldotriaconta-2,6,8,10,12,14,16,18,20,22,24,26,30-tridecaene
CC(=CCCC(=CC=CC(=CC=CC(=CC=CC=C(C)C=CC=C(C)C=CC=C(C)CCC=C(C)C)C)C)C)C
Color Red
Quantity 1 mg
Formula Weight 536.9
Percent Purity >90%
Physical Form Crystalline Needles
Chemical Name or Material Lycopene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'