missing translation for 'onlineSavingsMsg'
Learn More

Ly-49I Mouse anti-Mouse, PE, Clone: YLI-90, eBioscience™

Mouse Monoclonal Antibody

99.25 EUR - 164.00 EUR

Specifications

Antigen Ly-49I
Clone YLI-90
Target Species Mouse
Conjugate PE
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15278889 Affymetrix eBioscience
12-5895-81
50 μg 99.25 EUR
50µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 25-06-2019
Add to basket
 
15288889 Affymetrix eBioscience
12-5895-82
100 μg 164.00 EUR
100µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 25-06-2019
Add to basket
 
Description

Description

Ly49 receptors exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.
Specifications

Specifications

Ly-49I
Mouse
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
YLI-90
PE
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.