missing translation for 'onlineSavingsMsg'
Learn More

Luteolin-7-rutinoside, MedChemExpress

Product Code. 16462599
Encompass_Preferred
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 16462599

Brand: Medchem Express HYN6647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Luteolin-7-rutinoside has both anti-arthritic and antifungal activities, can result in a combination therapy for the treatment of fungal arthritis due to C. albicans infection.

Luteolin-7-rutinoside has both anti-arthritic and antifungal activities, can result in a combination therapy for the treatment of fungal arthritis due to C. albicans infection.

Specifications

Chemical Name or Material Luteolin-7-rutinoside
Physical Form Solid
CAS 20633-84-5
Color Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C27H30O15
For Use With (Application) COVID-19-immunoregulation
SMILES O=C1C=C(C2=CC=C(O)C(O)=C2)OC3=CC(O[C@H]4[C@@H]([C@H]([C@@H]([C@@H](CO[C@H]5[C@@H]([C@@H]([C@H]([C@H](C)O5)O)O)O)O4)O)O)O)=CC(O)=C13
Molecular Weight (g/mol) 594.52
Formula Weight 594.52
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.