Learn More
BRAND™ Low Form Glass Beaker
Shop All Brand Products
Click to view available options
Capacity (Metric):
5 mL
10 mL
50 mL
Unit Size:
Pack of 10
Description
- DIN EN ISO 3819
- Includes a spout
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Material | Borosilicate Glass |
| Includes | Spout |
| Type | Low Form |
| Graduated | Yes |