Learn More
Lovibond™ Reagent Set: Powder Pack; Oxygen Scavenger; DEHA
Premeasured unit dose reagent for water quality testing
Brand: Lovibond™ 536000
Additional Details : Weight : 0.30000kg Transport : UN number : 2031 Chem class : 8 Pack group : II
Includes: Oxyscav Reagent (10mL sample, 100 pack), DEHA 2 Reagent Solution (100mL)
Description
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Orbeco-Hellige and Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
10mL Oxyscav reagent (100 pack), 10mL DEHA 2 reagent solution | |
20 to 500μg | |
DEHA Reagent Set |
Hach™ and other manufacturers test kits, colorimeters and spectrophotometers | |
100 | |
PPST |
Safety and Handling
- 100TEST TESTS VARIO DEHA REAGENT SET
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : nitric acid, iron(III) nitrate nonahydrate