Learn More
Lovibond™ PD250 Reagent Dispenser Refill: DPD Free Chlorine
For use SC450 Chlorine and pH colorimeter, MC500 colorimeter and SP600 spectrophotometer
Brand:  Lovibond™ 530140
Additional Details : Weight : 0.30000kg
Product Code. 15331705
  • 90.30 EUR / Pack of 2
Estimated Shipment: 19-06-2024
Specifications
For chlorine testing
Includes 2 vial of DPD free chlorine reagent
Replacement Free Chlorine Reagent
Used with SC450 chlorine and pH colorimeter, MC500 colorimeter and SP600 spectrophotometer
250 Tests
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Hazard Statement
  • EUH210-Safety data sheet available on request.