Learn More
Lovibond™ Aqua Comparitor Color Discs: pH
For testing water with the Aqua Comparator
Brand:  Lovibond™ 146110
Additional Details : Weight : 0.02900kg
 View more versions of this product
Product Code. 11725448
  • 26.10 EUR / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for pH using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
Testing water with the Aqua comparator™
Aqua comparator™
5.2 to 6.8 pH
pH Bromcresol Purple Color Disc