Learn More
Lovibond™ Aqua Comparitor Color Discs: Iron
For testing water with the Aqua Comparator
Brand:  Lovibond™ 146220
Additional Details : Weight : 0.02900kg
 View more versions of this product
Product Code. 11775448
  • 26.10 EUR / Each
Estimated Shipment: 03-06-2024
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for iron using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
Testing water with the Aqua comparator™
Aqua comparator™
0.05 to 1mg/L
Iron HR Color Disc