Learn More
Lovibond™ Aqua Comparitor Color Discs: Copper
For testing water with the Aqua Comparator
Brand: Lovibond™ 146430
Additional Details : Weight : 0.02900kg
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for copper using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ | |
0 to 5mg/L | |
Copper HR Color Disc |