Learn More
Lovibond™ Aqua Comparitor Color Discs: Chlorine
For testing water with the Aqua Comparator
Brand: Lovibond™ 146330
Additional Details : Weight : 0.03000kg
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for chlorine using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ | |
0.01 to 0.2mg/L | |
Chlorine Dioxide Color Disc |