Learn More
SeaKem™ GTG™ Agarose, Lonza™
Specifically designed for preparative DNA electrophoresis. Lonza™ SeaKem™ GTG™ Agarose is a standard gelling temperature, high gel strength agarose that resolves DNA fragments greater than 1,000 bp.
Brand: Lonza 50070
Additional Details : Weight : 1.07000kg
Description
- DNA recovered from SeaKem™ GTG™ Agarose gels may be reliably digested and ligated
- Extensively performance tested to ensure complete compatibility with routine molecular biology techniques
Specifications
SeaKem GTG Agarose | |
125 g |