Learn More
Lonza™ BioWhittaker™ UltraMDCK™ Kidney Cell Media
Supports the growth of MADIN-DARBY Canine Kidney cells as well as other kidney-derived cell lines at low and high plating densities
Brand: Lonza™ BE12-749Q
Additional Details : Weight : 1.00000kg
Description
- Chemically-defined media optimized for specific cell types
- Low protein formulation
† Store as directed on label.
Specifications
2° to 8°C | |
Serum Free Medium (SFM) | |
1 L |