Learn More
Lonza™ BioWhittaker™ Trypsin Reagent
Used to remove cells from the culture vessel surface into suspension. BioWhittaker™ Trypsin Reagentis porcine parvovirus tested and mycoplasma screened
Brand: Lonza™ BE17-161F
Additional Details : Weight : 6.12350kg
Description
- Clear, colorless-yellow
- Porcine enzyme preparation, used to remove cells from the culture vessel surface into suspension. It is Generally used at a working concentration of 0.25%
- Prepared in a balanced salt solution, without calcium or magnesium at a pH of 7.6-7.8
- Trypsin activity can be halted by adding a serum media mixture or a Trypsin inhibitor
- Porcine parvovirus tested and mycoplasma screened
- Product is manufactured with irradiated trypsin
Specifications
-10° to -20°C | |
Sterile | |
500 mL |
Cell Culture Dissociation Reagent | |
7.1 to 7.9 |