Learn More
Lonza™ BioWhittaker™ Alpha MEM Eagle Medium
Used to culture bone marrow cells and amniotic cells for chromosome analysis. BioWhittaker™ Alpha MEM Eagle Medium are manufactured in accordance with cGMP regulations.
Brand:  Lonza™
Additional Details : Weight : 0.64000kg
Product Code. 11635190
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Differs from standard MEM by containing the following unique components:
  • Vitamin B12
  • Ascorbic acid
  • Non-essential amino acids
  • Sodium pyruvate
  • Lipoic acid
  • D-Biotin
Specifications
Specifications
2° to 8°C
Sterile-filtered through two consecutive 0.2 μm filters
6.91 to 7.27
Sterile
No Glutamine, No Deoxyribonucleosides, No Ribonucleosides
500 mL