missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ AccuGENE™ LB Broth
Ready-to-use solutions for wide range of molecular biology applications
Brand: Lonza™ 51217
Additional Details : Weight : 1.36080kg
Description
- Reagent prepeared with 18 megOhm water, filtered using 0.2μ filter, and filled into sterile bottles
Specifications
18° to 24°C | |
Switzerland | |
500 mL | |
Ideal for a wide range of molecular biology application | |
500 mL |
Liquid | |
LB Broth | |
Bacto Tryptone 10g/L, Bacto Yeast extract 5g/L, NaCl 10g/L | |
Bottle |