Learn More
Lonza™ AccuGENE™ 10% SDS (Monosodium Salt)
Product Code. 11508025
Click to view available options
Quantity:
500 mL
Unit Size:
Each
Product Code. 11508025
Brand: Lonza™ 51213
Ideal for a wide range of molecular biology applications
  • Ready-to-use
  • Prepared with 18megohm water
  • Filtered using 0.2μ filter and filled into sterile bottles
  • Manufactured according to strict quality control standards to ensure consistency
  • Guaranteed DNase, RNase and protease-free
  • Customized solutions are available to meet individual needs
TRUSTED_SUSTAINABILITY
Specifications
DNase Yes
Formulation 10% SDS (w/v), monosodium salt
For Use With (Application) Ideal for a wide range of molecular biology applications
Product Type 10% SDS (Monosodium Salt)
RNase Yes
Quantity 500 mL
For research or manufacturing use only.