Learn More
Lenz Laborglasintrumente Long Ground Sockets
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100607
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10449362
  • 155.82 EUR / Pack of 10
Estimated Shipment: 02-07-2024
Description
  • Conical ground sockets
  • Made of DURAN™ tubing
Specifications
Specifications
10/Pk.
1.5 mm
Borosilicate Glass (3.3)
NS 7/25
10 mm