missing translation for 'onlineSavingsMsg'
Learn More

Shamrock™ Litho Color Coded Dot Labels

Offered in a number of colors and diameters to fit your needs. Shamrock™ Color Coded Dot Labels offer multiple opportunities for color coding in the laboratory - test, physician or department.

Brand:  Shamrock™ SDL75R3

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11746985

  • 9.19 EUR / Pack of 1000


Estimated Shipment
08-03-2021
Add to basket

Description

Description

  • Resistant to water, oil and acid
  • Temperature range of -20 to +121°C
  • For autoclave, freezer, refrigerator and oven
  • Universal marking with ballpoint pen, paper pencil and felt
Specifications

Specifications

Color Coded Dot Label
Green
Litho
Permanent
19 mm
Videos
SDS
Documents