missing translation for 'onlineSavingsMsg'
Learn More

Shamrock™ Litho Color Coded Dot Labels

Offered in a number of colors and diameters to fit your needs. Shamrock™ Color Coded Dot Labels offer multiple opportunities for color coding in the laboratory - test, physician or department.

Brand:  Shamrock™ SDL-25R-4

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11756975

  • 11.95 EUR / Pack of 1000
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Resistant to water, oil, and acid
  • Temperature range of -20 to +121°C
  • For autoclave, freezer, refrigerator and oven
  • Universal marking with ballpoint pen, paper pencil and felt
Specifications

Specifications

Permanent
Roll
Color Coded Dot Label
Red
Litho
6.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers