missing translation for 'onlineSavingsMsg'
Learn More

Listeria monocytogenes p60 Antibody (p6007), DyLight 405, Novus Biologicals™

Product Code. 18215036 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215036 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215036 Supplier Novus Biologicals Supplier No. NBP280120V
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Listeria monocytogenes p60 Monoclonal specifically detects Listeria monocytogenes p60 in Bacteria samples. It is validated for ELISA/Immunocytochemistry/Immunofluorescence/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Listeria monocytogenes p60
Applications ELISA, Immunocytochemistry/Immunofluorescence, Western Blot
Classification Monoclonal
Clone p6007
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias IAP p60 from Listeria Monocytogenes, Invasion-associated Protein p60, Invasion-associated Protein p60 from Listeria Monocytogenes, invasion-associated-protein, p60 protein
Host Species Mouse
Immunogen Recombinant Listeria monocytogenes p60.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.