Learn More
MP Biomedicals™ Linbro™ Liquisystem, Cap-Bands
Used with combination of Tube 96 & I.D. Labels 96 to form a complete system for the storage of samples up to 1.0 mL is created. Once filled with samples, the tube can be sealed in an air tight fashion using the Cap Band 8.
Brand: MP Biomedicals™ 0961227B1
Additional Details : Weight : 0.01000kg
Specifications
Linbro Liquisystem | |
Used with combination of Tube 96 and I.D. Labels 96 to form a complete system for the storage of samples up to 1mL is created. | |
120/Cs. |