Learn More
Grant Instruments™ Plastic Lid for Grant Stirred Water Baths
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Plastic Lid for Grant Stirred Water Baths is constructed of curved or flat plastic with an integral handle
Brand: Grant Instruments™ PL5
Additional Details : Weight : 0.01000kg
Description
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Plastic Lid for Grant Stirred Water Baths is constructed of curved or flat plastic with an integral handle
- Flat or curved moulded plastic
- Integral handle
- For use with Grant Stirred Water Baths and heated Circulators
- Prevents heat from escaping
- Reduces sample contamination
Specifications
Lid for 5L tank | |
P5 Plastic Tank | |
Plastic |