missing translation for 'onlineSavingsMsg'
Learn More

Lead Standard, 1,000ug/mL in 2-5% HNO3 (Plasma Grade), SPEX CertiPrep™

Catalog Number p-218848
Click to view available options
Quantity:
250 mL
Packaging:
Poly Bottle
This item is not returnable. View return policy
7439-92-1, 7697-37-2, 7732-18-5
Pb
207.20
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
5352425
CHEBI:27889
lead
[Pb]
This item is not returnable. View return policy
7439-92-1, 7697-37-2, 7732-18-5
Pb
207.20
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
5352425
CHEBI:27889
lead
[Pb]

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Lead Standard
  • Made with ASTM Type I Water
  • 1,000 μg/mL concentration
  • 250 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

CAS Min % 1.0, 5.0, 94.0
CAS Max % 1.0, 5.0, 94.0
Grade Plasma
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
2-5% HNO3 1000μg/mL PLPB2-2Y, -2T, -2X
Packaging Poly Bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Quantity 250 mL
Chemical Name or Material Lead Standard
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'