Learn More
BRAND™ LDPE Wash Bottles with Interchangeable Wash Head
Catalog Number p-8004177
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Flexible LDPE construction. BRAND™ LDPE Wash Bottles with Interchangeable Wash Head includes an adjustable height delivery tube.
  • Flexible LDPE construction
  • Detachable thread wash head
  • Adjustable height delivery tube
Capacity (Metric) 500 mL
Material LDPE
No. per Pack 50/PK
Type Wash Bottle
Thread Size (Metric) GL-25
Diameter (Metric) Outer 75mm
Closure Included Yes
Graduated No
Height (Metric) 279 mm