Learn More
BRAND™ LDPE Dropping Bottles
Catalog Number 11823940
Click to view available options
Capacity (Metric):
20 mL
30 mL
50 mL
100 mL
250 mL
Flexible LDPE construction. BRAND™ LDPE Dropping Bottles include a detachable threaded cap with dropper nozzle and attached spout cap.
  • Flexible LDPE construction
  • Includes threaded cap with dropper nozzle and attached spout
Capacity (Metric) 30 mL
Height (Metric) With Closure 96mm
Material LDPE
Packaging 100/PK
Thread Size (Metric) GL 14
Diameter (Metric) Outer 34mm
Closure Included Yes
Closure Type Dropper Nozzle