Learn More
Thermo Scientific™ Large O-ring for Eutech™ ECDOGEN-S and ECDOTPII-S Dissolved Oxygen Electrodes
Thermo Scientific™ offers Large O-ring replacement parts for ECDOGEN-S and ECDOTPII-S Dissolved Oxygen Electrodes.
Brand:  Thermo Scientific™ 32X246702
Additional Details : Weight : 0.10000kg
Product Code. 11543875
  • 20.40 EUR / Each
Estimated Shipment: 18-04-2024
Description
Eutech™ Large O-ring is used for replacement parts in ECDOGEN-S and ECDOTPII-S Dissolved Oxygen Electrodes. These galvanic electrodes are designed to be rugged and long- lasting.
Features:
  • Replacement Part for Dissolved Oxygen Process Electrodes
Specifications
Specifications
ECDOGEN-S, ECDOTPII-S DO Electrode
Replacement Part