Learn More
Masterflex™ Large-bore Adapter with Male Luer to Hose Barb Connection
Larger bore for enhanced fluid flow
Brand: Masterflex™ 45509-50
Additional Details : Weight : 0.80000kg
Description
- Cole-Parmer offers a variety of barbed luer adapters to meet your application's needs.
- Standard hose barb for use with flexible tubing.
- Smooth bore hose barb to reduce flow turbulence and risk of line contamination.
- Wide-bore hose barb for enhanced fluid path flow.
- Low-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene.
- Manufactured in compliance with ISO 594-1 standards, these fittings are inspected to be free from contaminants, and are fully traceable.
- All parts meet tight dimensional tolerances for increased reliability and process purity.
- Choose from a range of sizes, 1⁄16 to 1⁄4 in. with a variety of connection options.
- Sold in packs of 25
Specifications
Barb x Luer | |
Polycarbonate | |
25/Pk. | |
3/16 in. |
Not Autoclavable | |
Large-bore adapter luer-style male x 3/16 in. hose barb, polycarbonate, pack of 25 | |
3/16 in. | |
Ethylene Oxide, Gamma |