Learn More
Masterflex™ Large-bore Adapter with Male Luer to Hose Barb Connection
Larger bore for enhanced fluid flow
Brand:  Masterflex™ 45509-50
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11729949
  • 43.93 EUR / Pack of 25
Estimated Shipment
04-03-2021
Add to basket
Description
Description
  • Cole-Parmer offers a variety of barbed luer adapters to meet your application's needs.
  • Standard hose barb for use with flexible tubing.
  • Smooth bore hose barb to reduce flow turbulence and risk of line contamination.
  • Wide-bore hose barb for enhanced fluid path flow.
  • Low-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene.
  • Manufactured in compliance with ISO 594-1 standards, these fittings are inspected to be free from contaminants, and are fully traceable.
  • All parts meet tight dimensional tolerances for increased reliability and process purity.
  • Choose from a range of sizes, 1⁄16 to 1⁄4 in. with a variety of connection options.
  • Sold in packs of 25
Specifications
Specifications
Barb x Luer
Polycarbonate
25/Pk.
3/16 in.
Not Autoclavable
Large-bore adapter luer-style male x 3/16 in. hose barb, polycarbonate, pack of 25
3/16 in.
Ethylene Oxide, Gamma