Learn More
Scientific Industries SI™ Large Ampule/Tube Attachment
Provides vigorous end-to-end tube and ampule agitation. Scientific Industries SI™ Large Ampule/Tube Attachment accommodates up to 10 to 17mm diameter tubes or ampules.
Brand: Scientific Industries SI™ SI-0511
Additional Details : Weight : 0.75000kg
Description
- Used with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse
- Tubes and ampules snap into place for vigorous end-to-end agitation
- Holds up to four 10 to 17mm diameter tubes or ampules
Specifications
Tubes | |
10 to 17 mm dia. | |
Vortex-Genie 2 family of mixers, Vortex-Genie Pulse | |
4 |