missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ Tb Emission Filters, 25 mm
LanthaScreen™ Tb Emission Filters, 25 mm
Brand: Invitrogen™ PV00325
Additional Details : Weight : 0.03000kg
Description
A set of two emission filters (diameter = 25 mm) for use with the LanthaScreen™ Tb assays and products: 495 nm filter with a 10 nm bandpass, and a 520 nm filter with a 25 nm bandwidth. For use primarily with the Molecular Devices Analyst plate reader.For assistance in setting up your instrument, please contact our Technical Support team in North America at +1 760 603 7200, x40266.
Specifications
Tb Emission Filters | |
Microplate Reader | |
495/10, 520/25 nm | |
Pharma and biopharma, target and lead identification and validation |
For Research Use Only. Not for use in diagnostic procedures.